Assay Details
| Gene Symbol: FAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105221.3 | NP_176725.1 | 4 - 5 | 1041 | 83 | |
| GenBank mRNA | AY126994.1 | - | 4 - 5 | 990 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAS1| Gene Symbol: FAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105221.3 | NP_176725.1 | 4 - 5 | 1041 | 83 | |
| GenBank mRNA | AY126994.1 | - | 4 - 5 | 990 | 83 | |
Set Membership: |
|