Assay Details
| Gene Symbol: FAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198391.1 | NP_001185320.1 | 8 - 9 | 1879 | 73 | |
| NM_105221.3 | NP_176725.1 | 8 - 9 | 1930 | 73 | ||
| GenBank mRNA | AY126994.1 | - | 8 - 9 | 1879 | 73 | |
More Information
Set Membership: |
|

FAS1
Genomic Map