Assay Details
Gene Symbol: FRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334216.1 | NP_001321558.1 | 2 - 3 | 1131 | 96 | |
NM_105232.3 | NP_176736.2 | 2 - 3 | 1114 | 96 | ||
GenBank mRNA | AY761186.1 | - | 2 - 3 | 1087 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
