Assay Details
Gene Symbol: FRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334216.1 | NP_001321558.1 | 7 - 8 | 2364 | 72 | |
NM_105232.3 | NP_176736.2 | 7 - 8 | 2347 | 72 | ||
GenBank mRNA | AY761186.1 | - | 7 - 8 | 2320 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
