Assay Details
Gene Symbol: UCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105238.3 | NP_564858.1 | 4 - 5 | 319 | 99 | |
GenBank mRNA | AY065083.1 | - | 4 - 5 | 242 | 99 | |
AY114549.1 | - | 3 - 4 | 120 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
