Assay Details
| Gene Symbol: LSM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036161.1 | NP_001031238.1 | 2 - 3 | 135 | 68 | |
| NM_001198394.1 | NP_001185323.1 | 2 - 3 | 114 | 68 | ||
| NM_105244.4 | NP_176747.1 | 2 - 3 | 345 | 68 | ||
| GenBank mRNA | AY070767.1 | - | 2 - 3 | 168 | 68 | |
| AY094059.1 | - | 1 - 2 | 37 | 68 | ||
More Information
Set Membership: |
|

LSM8
Genomic Map