Assay Details
| Gene Symbol: GSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036163.2 | NP_001031240.1 | 2 - 3 | 233 | 73 | |
| NM_001198398.1 | NP_001185327.1 | 2 - 3 | 278 | 73 | ||
| NM_105291.4 | NP_176794.1 | 2 - 3 | 298 | 73 | ||
| GenBank mRNA | AY091101.1 | - | 2 - 3 | 206 | 73 | |
| AY122962.1 | - | 2 - 3 | 115 | 73 | ||
More Information
Set Membership: |
|

GSR2
Genomic Map