Assay Details
| Gene Symbol: ETR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105305.4 | NP_176808.3 | 2 - 3 | 1073 | 82 | |
| Gene Symbol: AT2G45180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230334.1 | - | 1277 | 82 | ||
More Information
Set Membership: |
|

ETR1
Genomic Map