Assay Details
| Gene Symbol: GCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036170.1 | NP_001031247.1 | 14 - 15 | 1663 | 136 | |
| NM_105416.5 | NP_176916.2 | 15 - 16 | 2053 | 136 | ||
More Information
Set Membership: |
|

GCS1
Genomic Map