Assay Details
Gene Symbol: GCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036170.1 | NP_001031247.1 | 8 - 9 | 1015 | 123 | |
NM_105416.5 | NP_176916.2 | 9 - 10 | 1405 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
