Assay Details
| Gene Symbol: LOX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105423.3 | NP_176923.1 | 7 - 8 | 1850 | 74 | |
| GenBank mRNA | AJ748537.1 | - | 7 - 8 | 1728 | 74 | |
| AY081253.1 | - | 7 - 8 | 1755 | 74 | ||
| BT010546.1 | - | 7 - 8 | 1728 | 74 | ||
More Information
Set Membership: |
|

LOX6
Genomic Map