Assay Details
| Gene Symbol: LOX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105423.3 | NP_176923.1 | 8 - 9 | 2117 | 80 | |
| GenBank mRNA | AJ748537.1 | - | 8 - 9 | 1995 | 80 | |
| AY081253.1 | - | 8 - 9 | 2022 | 80 | ||
| BT010546.1 | - | 8 - 9 | 1995 | 80 | ||
More Information
Set Membership: |
|

LOX6
Genomic Map