Assay Details
| Gene Symbol: ARR11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334322.1 | NP_001320474.1 | 1 - 2 | 428 | 83 | |
| NM_105439.3 | NP_176938.1 | 1 - 2 | 428 | 83 | ||
| GenBank mRNA | AK228216.1 | - | 1 - 2 | 207 | 83 | |
| BT008768.1 | - | 1 - 2 | 116 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001334322.1 transcripts.
Set Membership: |
|

ARR11
Genomic Map