Assay Details
| Gene Symbol: ARR11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334321.1 | NP_001320475.1 | 2 - 3 | 1041 | 87 | |
| NM_001334322.1 | NP_001320474.1 | 2 - 3 | 1034 | 87 | ||
| NM_105439.3 | NP_176938.1 | 3 - 4 | 960 | 87 | ||
| GenBank mRNA | AK228216.1 | - | 3 - 4 | 739 | 87 | |
| BT008768.1 | - | 3 - 4 | 648 | 87 | ||
More Information
Set Membership: |
|

ARR11
Genomic Map