Assay Details
| Gene Symbol: ARR11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334321.1 | NP_001320475.1 | 3 - 4 | 1118 | 87 | |
| NM_001334322.1 | NP_001320474.1 | 3 - 4 | 1111 | 87 | ||
| NM_105439.3 | NP_176938.1 | 4 - 5 | 1037 | 87 | ||
| GenBank mRNA | AK228216.1 | - | 4 - 5 | 816 | 87 | |
| BT008768.1 | - | 4 - 5 | 725 | 87 | ||
More Information
Set Membership: |
|

ARR11
Genomic Map