Assay Details
| Gene Symbol: TEL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105445.2 | NP_176943.1 | 4 - 5 | 1490 | 102 | |
| GenBank mRNA | DQ446408.1 | - | 4 - 5 | 1311 | 102 | |
More Information
Set Membership: |
|

TEL2
Genomic Map