Assay Details
| Gene Symbol: BBX27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334352.1 | NP_001321976.1 | 5 - 6 | 1562 | 78 | |
| NM_001334353.1 | NP_001321977.1 | 4 - 5 | 1581 | 78 | ||
| NM_105490.4 | NP_176986.1 | 5 - 6 | 1111 | 78 | ||
More Information
Set Membership: |
|

BBX27
Genomic Map