Assay Details
Gene Symbol: PAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105536.4 | NP_177031.1 | 7 - 8 | 983 | 79 | |
GenBank mRNA | AK118286.1 | - | 7 - 8 | 804 | 79 | |
BT008586.1 | - | 7 - 8 | 636 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
