Assay Details
Gene Symbol: PAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105536.4 | NP_177031.1 | 9 - 10 | 1319 | 103 | |
GenBank mRNA | AK118286.1 | - | 9 - 10 | 1140 | 103 | |
BT008586.1 | - | 9 - 10 | 972 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
