Assay Details
| Gene Symbol: TADA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105544.3 | NP_177039.1 | 3 - 4 | 3816 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TADA| Gene Symbol: TADA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105544.3 | NP_177039.1 | 3 - 4 | 3816 | 78 | |
Set Membership: |
|