Assay Details
Gene Symbol: PHO1;H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334380.1 | NP_001322237.1 | 10 - 11 | 2807 | 87 | |
NM_105547.3 | NP_564940.1 | 11 - 12 | 2669 | 87 | ||
GenBank mRNA | AY057574.1 | - | 11 - 12 | 2343 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
