Assay Details
Gene Symbol: MGP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198426.1 | NP_001185355.1 | 9 - 10 | 2222 | 79 | |
NM_105571.4 | NP_177063.1 | 9 - 10 | 2272 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
