Assay Details
Gene Symbol: BIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105572.5 | NP_177064.1 | 3 - 4 | 433 | 152 | |
GenBank mRNA | BT002352.1 | - | 3 - 4 | 258 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
