Assay Details
| Gene Symbol: SIK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105589.4 | NP_564955.1 | 9 - 10 | 1525 | 64 | |
| NM_179534.2 | NP_849865.1 | 9 - 10 | 1330 | 64 | ||
| GenBank mRNA | AY072199.1 | - | 9 - 10 | 1330 | 64 | |
| AY133857.1 | - | 9 - 10 | 1278 | 64 | ||
| U96613.1 | - | 9 - 10 | 1363 | 64 | ||
More Information
Set Membership: |
|

SIK1
Genomic Map