Assay Details
Gene Symbol: AFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105593.5 | NP_564956.1 | 1 - 2 | 1084 | 67 | |
GenBank mRNA | AY136357.1 | - | 1 - 2 | 828 | 67 | |
BT000199.1 | - | 1 - 2 | 675 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
