Assay Details
Gene Symbol: AGO7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105611.5 | NP_177103.1 | 1 - 2 | 655 | 66 | |
GenBank mRNA | AY394564.1 | - | 1 - 2 | 493 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
