Assay Details
| Gene Symbol: AGL94 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334413.1 | NP_001322116.1 | 2 - 3 | 604 | 82 | |
| NM_105623.5 | NP_177113.3 | 2 - 3 | 604 | 82 | ||
| GenBank mRNA | AB115013.1 | - | 1 - 2 | 181 | 82 | |
More Information
Set Membership: |
|

AGL94
Genomic Map