Assay Details
| Gene Symbol: AGL94 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334413.1 | NP_001322116.1 | 8 - 9 | 925 | 104 | |
| NM_001334414.1 | NP_001322115.1 | 4 - 5 | 285 | 104 | ||
| NM_105623.5 | NP_177113.3 | 8 - 9 | 925 | 104 | ||
| GenBank mRNA | AB115013.1 | - | 7 - 8 | 502 | 104 | |
More Information
Set Membership: |
|

AGL94
Genomic Map