Assay Details
| Gene Symbol: RPL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105630.4 | NP_177120.1 | 1 - 2 | 205 | 62 | |
| GenBank mRNA | AF324703.1 | - | 1 - 2 | 142 | 62 | |
| AF327531.1 | - | 1 - 2 | 141 | 62 | ||
| AF349526.1 | - | 1 - 2 | 80 | 62 | ||
| AF446885.1 | - | 1 - 2 | 80 | 62 | ||
| AY052720.1 | - | 1 - 2 | 142 | 62 | ||
| AY085544.1 | - | 1 - 2 | 142 | 62 | ||
More Information
Set Membership: |
|

RPL34
Genomic Map