Assay Details
| Gene Symbol: RPL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105630.4 | NP_177120.1 | 3 - 4 | 388 | 92 | |
| GenBank mRNA | AF324703.1 | - | 3 - 4 | 325 | 92 | |
| AF327531.1 | - | 3 - 4 | 324 | 92 | ||
| AF349526.1 | - | 3 - 4 | 263 | 92 | ||
| AF446885.1 | - | 3 - 4 | 263 | 92 | ||
| AY052720.1 | - | 3 - 4 | 325 | 92 | ||
| AY085544.1 | - | 3 - 4 | 325 | 92 | ||
More Information
Set Membership: |
|

RPL34
Genomic Map