Assay Details
Gene Symbol: HO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001124102.1 | NP_001117574.1 | 4 - 5 | 866 | 109 | |
NM_001334420.1 | NP_001321799.1 | 3 - 4 | 854 | 109 | ||
NM_105640.4 | NP_177130.1 | 4 - 5 | 993 | 109 | ||
GenBank mRNA | AY084250.1 | - | 4 - 5 | 858 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
