Assay Details
| Gene Symbol: HEMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084331.3 | NP_001077800.1 | 12 - 13 | 1337 | 79 | |
| NM_001334421.1 | NP_001320950.1 | 11 - 12 | 1451 | 79 | ||
| NM_105642.4 | NP_177132.1 | 11 - 12 | 1363 | 79 | ||
| GenBank mRNA | AF327428.1 | - | 11 - 12 | 1071 | 79 | |
| AF361803.1 | - | 11 - 12 | 1107 | 79 | ||
| AY059154.1 | - | 10 - 11 | 1042 | 79 | ||
| AY081254.1 | - | 11 - 12 | 1113 | 79 | ||
| AY113941.1 | - | 10 - 11 | 1042 | 79 | ||
| AY120705.1 | - | 11 - 12 | 1071 | 79 | ||
| AY128711.1 | - | 10 - 11 | 1042 | 79 | ||
| Gene Symbol: RAB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK230120.1 | - | 37 | 79 | ||
More Information
Set Membership: |
|

HEMB1
Genomic Map