Assay Details
| Gene Symbol: HEMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084331.3 | NP_001077800.1 | 5 - 6 | 791 | 85 | |
| NM_001334421.1 | NP_001320950.1 | 4 - 5 | 905 | 85 | ||
| NM_105642.4 | NP_177132.1 | 4 - 5 | 817 | 85 | ||
| GenBank mRNA | AF327428.1 | - | 4 - 5 | 525 | 85 | |
| AF361803.1 | - | 4 - 5 | 561 | 85 | ||
| AY059154.1 | - | 3 - 4 | 496 | 85 | ||
| AY081254.1 | - | 4 - 5 | 567 | 85 | ||
| AY113941.1 | - | 3 - 4 | 496 | 85 | ||
| AY120705.1 | - | 4 - 5 | 525 | 85 | ||
| AY128711.1 | - | 3 - 4 | 496 | 85 | ||
More Information
Set Membership: |
|

HEMB1
Genomic Map