Assay Details
| Gene Symbol: HEMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084331.3 | NP_001077800.1 | 7 - 8 | 940 | 98 | |
| NM_001334421.1 | NP_001320950.1 | 6 - 7 | 1054 | 98 | ||
| NM_105642.4 | NP_177132.1 | 6 - 7 | 966 | 98 | ||
| GenBank mRNA | AF327428.1 | - | 6 - 7 | 674 | 98 | |
| AF361803.1 | - | 6 - 7 | 710 | 98 | ||
| AY059154.1 | - | 5 - 6 | 645 | 98 | ||
| AY081254.1 | - | 6 - 7 | 716 | 98 | ||
| AY113941.1 | - | 5 - 6 | 645 | 98 | ||
| AY120705.1 | - | 6 - 7 | 674 | 98 | ||
| AY128711.1 | - | 5 - 6 | 645 | 98 | ||
More Information
Set Membership: |
|

HEMB1
Genomic Map