Assay Details
| Gene Symbol: HEMB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084331.3 | NP_001077800.1 | 9 - 10 | 1063 | 119 | |
| NM_001334421.1 | NP_001320950.1 | 8 - 9 | 1177 | 119 | ||
| NM_105642.4 | NP_177132.1 | 8 - 9 | 1089 | 119 | ||
| GenBank mRNA | AF327428.1 | - | 8 - 9 | 797 | 119 | |
| AF361803.1 | - | 8 - 9 | 833 | 119 | ||
| AY059154.1 | - | 7 - 8 | 768 | 119 | ||
| AY081254.1 | - | 8 - 9 | 839 | 119 | ||
| AY113941.1 | - | 7 - 8 | 768 | 119 | ||
| AY120705.1 | - | 8 - 9 | 797 | 119 | ||
| AY128711.1 | - | 7 - 8 | 768 | 119 | ||
More Information
Set Membership: |
|

HEMB1
Genomic Map