Assay Details
| Gene Symbol: AMY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105651.5 | NP_564977.1 | 11 - 12 | 2503 | 122 | |
| GenBank mRNA | AY050398.1 | - | 11 - 12 | 2379 | 122 | |
| BT000643.1 | - | 11 - 12 | 2333 | 122 | ||
More Information
Set Membership: |
|

AMY3
Genomic Map