Assay Details
| Gene Symbol: AMY3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105651.5 | NP_564977.1 | 12 - 13 | 2672 | 81 | |
| GenBank mRNA | AY050398.1 | - | 12 - 13 | 2548 | 81 | |
| BT000643.1 | - | 12 - 13 | 2502 | 81 | ||
More Information
Set Membership: |
|

AMY3
Genomic Map