Assay Details
| Gene Symbol: TH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105656.3 | NP_177146.1 | 2 - 3 | 451 | 70 | |
| GenBank mRNA | AK117825.1 | - | 2 - 3 | 370 | 70 | |
| BT003670.1 | - | 2 - 3 | 309 | 70 | ||
More Information
Set Membership: |
|

TH8
Genomic Map