Assay Details
| Gene Symbol: BCA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105709.4 | NP_177198.1 | 2 - 3 | 410 | 59 | |
| GenBank mRNA | AY034926.1 | - | 2 - 3 | 192 | 59 | |
| AY113922.1 | - | 1 - 2 | 54 | 59 | ||
More Information
Set Membership: |
|

BCA4
Genomic Map