Assay Details
| Gene Symbol: BCA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105709.4 | NP_177198.1 | 6 - 7 | 776 | 77 | |
| NM_179541.3 | NP_849872.1 | 4 - 5 | 1275 | 77 | ||
| NM_202390.3 | NP_974119.1 | 5 - 6 | 1185 | 77 | ||
| GenBank mRNA | AY034926.1 | - | 6 - 7 | 558 | 77 | |
| AY087827.1 | - | 6 - 7 | 574 | 77 | ||
| AY113922.1 | - | 5 - 6 | 420 | 77 | ||
More Information
Set Membership: |
|

BCA4
Genomic Map