Assay Details
| Gene Symbol: AT1G70420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105710.3 | NP_564988.1 | 1 | 997 | 154 | |
| GenBank mRNA | AY045589.1 | - | 1 | 941 | 154 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G70420| Gene Symbol: AT1G70420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105710.3 | NP_564988.1 | 1 | 997 | 154 | |
| GenBank mRNA | AY045589.1 | - | 1 | 941 | 154 | |
Set Membership: |
|