Assay Details
| Gene Symbol: AOAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036185.1 | NP_001031262.1 | 4 - 5 | 445 | 85 | |
| NM_001036186.2 | NP_001031263.1 | 4 - 5 | 557 | 85 | ||
| NM_105726.3 | NP_177215.1 | 4 - 5 | 783 | 85 | ||
| NM_202393.2 | NP_974122.1 | 4 - 5 | 752 | 85 | ||
| GenBank mRNA | AF479640.1 | - | 3 - 4 | 317 | 85 | |
| AK316788.1 | - | 4 - 5 | 384 | 85 | ||
| AY035130.1 | - | 4 - 5 | 423 | 85 | ||
| AY062982.1 | - | 3 - 4 | 298 | 85 | ||
More Information
Set Membership: |
|

AOAT2
Genomic Map