Assay Details
| Gene Symbol: AOAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036185.1 | NP_001031262.1 | 8 - 9 | 948 | 91 | |
| NM_001036186.2 | NP_001031263.1 | 8 - 9 | 1060 | 91 | ||
| NM_105726.3 | NP_177215.1 | 8 - 9 | 1286 | 91 | ||
| NM_202393.2 | NP_974122.1 | 8 - 9 | 1255 | 91 | ||
| GenBank mRNA | AF479640.1 | - | 7 - 8 | 820 | 91 | |
| AK316788.1 | - | 8 - 9 | 887 | 91 | ||
| AY035130.1 | - | 8 - 9 | 926 | 91 | ||
| AY062982.1 | - | 7 - 8 | 801 | 91 | ||
More Information
Set Membership: |
|

AOAT2
Genomic Map