Assay Details
| Gene Symbol: ABCB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334463.1 | NP_001321943.1 | 15 - 16 | 2033 | 63 | |
| NM_105729.6 | NP_177218.3 | 15 - 16 | 2082 | 63 | ||
| GenBank mRNA | AK228750.1 | - | 15 - 16 | 2020 | 63 | |
| AY078409.1 | - | 15 - 16 | 1895 | 63 | ||
More Information
Set Membership: |
|

ABCB26
Genomic Map