Assay Details
Gene Symbol: SAG18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105788.4 | NP_177275.1 | 1 - 2 | 202 | 76 | |
GenBank mRNA | BT044602.1 | - | 1 - 2 | 124 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
