Assay Details
Gene Symbol: SAG18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198440.1 | NP_001185369.1 | 5 - 6 | 657 | 74 | |
NM_105788.4 | NP_177275.1 | 6 - 7 | 830 | 74 | ||
GenBank mRNA | BT044602.1 | - | 6 - 7 | 752 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
