Assay Details
Gene Symbol: EBS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198441.1 | NP_001185370.1 | 3 - 4 | 691 | 94 | |
NM_001334490.1 | NP_001323059.1 | 3 - 4 | 771 | 94 | ||
NM_105791.5 | NP_177278.3 | 3 - 4 | 691 | 94 | ||
GenBank mRNA | AK230327.1 | - | 3 - 4 | 691 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
