Assay Details
Gene Symbol: POK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105796.3 | NP_565015.1 | 17 - 18 | 1840 | 150 | |
GenBank mRNA | AF367286.1 | - | 17 - 18 | 1832 | 150 | |
BT004525.1 | - | 17 - 18 | 1691 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
