Assay Details
| Gene Symbol: POK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105796.3 | NP_565015.1 | 18 - 19 | 1912 | 154 | |
| GenBank mRNA | AF367286.1 | - | 18 - 19 | 1904 | 154 | |
| BT004525.1 | - | 18 - 19 | 1763 | 154 | ||
More Information
Set Membership: |
|

POK
Genomic Map