Assay Details
| Gene Symbol: POK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_105796.3 | NP_565015.1 | 4 - 5 | 417 | 84 | |
| GenBank mRNA | AF367286.1 | - | 4 - 5 | 409 | 84 | |
| BT004525.1 | - | 4 - 5 | 268 | 84 | ||
More Information
Set Membership: |
|

POK
Genomic Map